Product Details
- SNP ID
-
rs671445
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:78064621 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATATGCACATATATGTGTTTGTGCA[C/T]GTGTGCATTTATTAGTATGTGAATG
- Phenotype
-
MIM: 603845
MIM: 601926
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NDUFC2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs145855011] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NDUFC2
- Gene Name
- NADH:ubiquinone oxidoreductase subunit C2
There are no transcripts associated with this gene.
- Gene
- NDUFC2-KCTD14
- Gene Name
- NDUFC2-KCTD14 readthrough
- Gene
- THRSP
- Gene Name
- thyroid hormone responsive
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003251.3 |
|
Intron |
|
|
NP_003242.1 |
View Full Product Details