Product Details

SNP ID
rs986096
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:82733184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCGGCGCAGCCACCTCACCCCC[A/C]GCCGGCTCGGCACCCCCGGGGACGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM181B PubMed Links

Gene Details

Gene
FAM181B
Gene Name
family with sequence similarity 181 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175885.3 681 Silent Mutation GCG,GCT A182A NP_787081.2

View Full Product Details