Product Details

SNP ID
rs1063610
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:12583940 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCCCAATGCACTGGACACCTTA[A/G]AAGCTGTGAAAGGAGGACGTGTCCC
Phenotype
MIM: 608426 MIM: 164760
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MKRN2 PubMed Links

Gene Details

Gene
MKRN2
Gene Name
makorin ring finger protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271707.1 2936 Intron NP_001258636.1
NM_014160.4 2936 Intron NP_054879.3
Gene
RAF1
Gene Name
Raf-1 proto-oncogene, serine/threonine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002880.3 2936 UTR 3 NP_002871.1
XM_005265355.2 2936 UTR 3 XP_005265412.1
XM_005265357.2 2936 UTR 3 XP_005265414.1
XM_005265358.4 2936 UTR 3 XP_005265415.1
XM_005265359.4 2936 UTR 3 XP_005265416.1
XM_011533974.2 2936 UTR 3 XP_011532276.1
XM_011533975.2 2936 UTR 3 XP_011532277.1
XM_017006966.1 2936 UTR 3 XP_016862455.1

View Full Product Details