Product Details

SNP ID
rs1050592
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:39265293 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTCATTCTTCAAATTTTGAGCACA[A/G]TTCTCAACAACACTCTAGGGTTGTT
Phenotype
MIM: 601470
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CX3CR1 PubMed Links

Gene Details

Gene
CX3CR1
Gene Name
C-X3-C motif chemokine receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171171.1 1309 UTR 3 NP_001164642.1
NM_001171172.1 1309 UTR 3 NP_001164643.1
NM_001171174.1 1309 UTR 3 NP_001164645.1
NM_001337.3 1309 UTR 3 NP_001328.1

View Full Product Details