Product Details
- SNP ID
-
rs1105418
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:68482778 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATCCTTTTCCATCTTCTGTTGTTA[C/G]TGGTTTTGTGTCTCCTTTCTTGATT
- Phenotype
-
MIM: 610879
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PPP6R3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76865366] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PPP6R3
- Gene Name
- protein phosphatase 6 regulatory subunit 3
View Full Product Details