Product Details
- SNP ID
-
rs1417943
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:68962356 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGCTTCTTAAACAAGTGGTTAAAG[C/T]TTTTTCAGGCCCTCTGTGGTGGCTC
- Phenotype
-
MIM: 606357
MIM: 610373
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DDX21
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76725293] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DDX21
- Gene Name
- DEAD-box helicase 21
- Gene
- DDX50
- Gene Name
- DEAD-box helicase 50
There are no transcripts associated with this gene.
View Full Product Details