Product Details

SNP ID
rs1533133
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:25382683 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGCAAAGTTGCATGTGATGGTGATA[C/T]GCAGTGAAACTCACTGCCCTCTTTT
Phenotype
MIM: 606947 MIM: 611792
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANAPC4 PubMed Links

Gene Details

Gene
ANAPC4
Gene Name
anaphase promoting complex subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286756.1 Intron NP_001273685.1
NM_013367.2 Intron NP_037499.2
XM_005248159.2 Intron XP_005248216.1
XM_011513838.1 Intron XP_011512140.1
XM_011513839.1 Intron XP_011512141.1
XM_017008138.1 Intron XP_016863627.1
Gene
ZCCHC4
Gene Name
zinc finger CCHC-type containing 4
There are no transcripts associated with this gene.

View Full Product Details