Product Details

SNP ID
rs1061425
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:185916887 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGTTCATCCATGACAGGTATAG[G/T]GTTCCCTTATCATGTACTTTGAAGC
Phenotype
MIM: 602719
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TRA2B PubMed Links

Gene Details

Gene
TRA2B
Gene Name
transformer 2 beta homolog (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243879.1 4941 UTR 3 NP_001230808.1
NM_004593.2 4941 UTR 3 NP_004584.1
XM_005247703.1 4941 UTR 3 XP_005247760.1
XM_011513072.2 4941 UTR 3 XP_011511374.1
XM_017007033.1 4941 UTR 3 XP_016862522.1

View Full Product Details