Product Details

SNP ID
rs1652094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:49835657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATTTTTGTCTTTGTGTAAAACA[A/G]GTTTTTATCACAATTTTTTAAGTCC
Phenotype
MIM: 601193
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRISP1 PubMed Links

Gene Details

Gene
CRISP1
Gene Name
cysteine rich secretory protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001131.2 Intron NP_001122.2
NM_001205220.1 Intron NP_001192149.1
NM_170609.1 Intron NP_733758.1
XM_011514315.2 Intron XP_011512617.1
XM_017010320.1 Intron XP_016865809.1
XM_017010321.1 Intron XP_016865810.1

View Full Product Details