Product Details

SNP ID
rs1507713
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:58140 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCCTCACCAGATTTGGTCTCTCAA[C/T]GTTGGACTTCTCAGCCTCCACAACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLEKHG4B PubMed Links

Gene Details

Gene
PLEKHG4B
Gene Name
pleckstrin homology and RhoGEF domain containing G4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052909.3 Intron NP_443141.3
XM_006714446.3 Intron XP_006714509.1
XM_011513968.2 Intron XP_011512270.1
XM_011513969.2 Intron XP_011512271.1
XM_011513970.1 Intron XP_011512272.1
XM_011513971.2 Intron XP_011512273.1
XM_011513972.2 Intron XP_011512274.1
XM_011513973.1 Intron XP_011512275.1
XM_017009112.1 Intron XP_016864601.1

View Full Product Details