Product Details

SNP ID
rs277628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:100336302 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACTCTGGAGCAGAGGTGGGATGGG[A/G]AGTTCGGGCAGGCCATTACTAATTG
Phenotype
MIM: 616769
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NIT2 PubMed Links

Gene Details

Gene
NIT2
Gene Name
nitrilase family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020202.4 Intron NP_064587.1

View Full Product Details