Product Details
- SNP ID
-
rs839246
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:58013380 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GGTCACTAGGCAGCTTCCAGGGTGG[C/T]GGGTCACTGGTCCAGTCAGCTCCTT
- Phenotype
-
MIM: 603381
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FLNB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs149458554] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FLNB
- Gene Name
- filamin B
View Full Product Details