Product Details

SNP ID
rs1338508
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:115941933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAGTTACAGGATATTTTAAAAGA[A/G]AAAAAAAAATCTCAAAGCACAGGTC
Phenotype
MIM: 606573
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FRK PubMed Links

Gene Details

Gene
FRK
Gene Name
fyn related Src family tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002031.2 2636 UTR 3 NP_002022.1
XM_005266880.4 2636 UTR 3 XP_005266937.1
XM_005266881.2 2636 UTR 3 XP_005266938.1
XM_005266882.4 2636 UTR 3 XP_005266939.1
XM_011535653.2 2636 UTR 3 XP_011533955.1
XM_011535654.2 2636 UTR 3 XP_011533956.1
XM_011535655.2 2636 UTR 3 XP_011533957.1
XM_011535656.2 2636 UTR 3 XP_011533958.1
XM_017010645.1 2636 UTR 3 XP_016866134.1

View Full Product Details