Product Details

SNP ID
rs11361
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:38406593 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTGACATCATGACATCTACAAAAAC[A/G]GTAGAAAAACCCAGGTTTGGTGCGG
Phenotype
MIM: 100670 MIM: 610413
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDH1B1 PubMed Links

Gene Details

Gene
ALDH1B1
Gene Name
aldehyde dehydrogenase 1 family member B1
There are no transcripts associated with this gene.

Gene
IGFBPL1
Gene Name
insulin like growth factor binding protein like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007563.2 3494 UTR 3 NP_001007564.1
XM_017014699.1 3494 Intron XP_016870188.1

View Full Product Details