Product Details

SNP ID
rs940714
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:42897953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGGGAGGGGGAGGGATGCGCAGA[C/G]CCCGGGCGGACCCCGCGCTGCCTTC
Phenotype
MIM: 607825 MIM: 614649
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HOOK3 PubMed Links

Gene Details

Gene
HOOK3
Gene Name
hook microtubule-tethering protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032410.3 Intron NP_115786.1
Gene
MIR4469
Gene Name
microRNA 4469
There are no transcripts associated with this gene.

Gene
RNF170
Gene Name
ring finger protein 170
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160223.1 Intron NP_001153695.1
NM_001160224.1 Intron NP_001153696.1
NM_001160225.1 Intron NP_001153697.1
NM_030954.3 Intron NP_112216.3
XM_006716404.2 Intron XP_006716467.1
XM_006716405.2 Intron XP_006716468.1
XM_011544666.2 Intron XP_011542968.1
XM_017013881.1 Intron XP_016869370.1
XM_017013882.1 Intron XP_016869371.1

View Full Product Details