Product Details

SNP ID
rs7581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:109016967 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTTTTCCTATAAACCCTGAGGT[C/T]GAAAGCTCTGGATAGCTCACCTAAA
Phenotype
MIM: 604785
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTNNAL1 PubMed Links

Gene Details

Gene
CTNNAL1
Gene Name
catenin alpha like 1
There are no transcripts associated with this gene.

Gene
TMEM245
Gene Name
transmembrane protein 245
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032012.3 6048 UTR 3 NP_114401.2
XM_011518446.2 6048 UTR 3 XP_011516748.1
XM_011518447.1 6048 UTR 3 XP_011516749.1
XM_011518449.1 6048 UTR 3 XP_011516751.1
XM_011518451.1 6048 UTR 3 XP_011516753.1
XM_011518452.1 6048 UTR 3 XP_011516754.1
XM_017014571.1 6048 UTR 3 XP_016870060.1
XM_017014572.1 6048 UTR 3 XP_016870061.1

View Full Product Details