Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001318872.1 | Intron | NP_001305801.1 | |||
NM_006567.4 | Intron | NP_006558.1 | |||
XM_005248812.3 | Intron | XP_005248869.1 | |||
XM_006714966.2 | Intron | XP_006715029.1 | |||
XM_011514247.2 | Intron | XP_011512549.1 | |||
XM_011514248.2 | Intron | XP_011512550.1 | |||
XM_011514249.1 | Intron | XP_011512551.1 | |||
XM_011514251.2 | Intron | XP_011512553.1 | |||
XM_017010186.1 | Intron | XP_016865675.1 | |||
XM_017010187.1 | Intron | XP_016865676.1 |