Product Details

SNP ID
rs1128150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:113165577 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTTCAAAGTCATTGACCTCTTGT[A/G]TCAGATTTAAGGCAAAGAAGAAGAT
Phenotype
MIM: 603088
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FKBP15 PubMed Links

Gene Details

Gene
FKBP15
Gene Name
FK506 binding protein 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015258.1 4127 UTR 3 NP_056073.1
XM_006717018.2 4127 UTR 3 XP_006717081.1
XM_006717019.1 4127 UTR 3 XP_006717082.1
XM_017014552.1 4127 UTR 3 XP_016870041.1
Gene
SLC31A2
Gene Name
solute carrier family 31 member 2
There are no transcripts associated with this gene.

View Full Product Details