Product Details

SNP ID
rs1048550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:150264812 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTACAAATTGCTAATCAAAAGGAA[A/G]GTGGAGATGATAGTTTAATAACAAA
Phenotype
MIM: 613166 MIM: 606453
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCLK2 PubMed Links

Gene Details

Gene
DCLK2
Gene Name
doublecortin like kinase 2
There are no transcripts associated with this gene.

Gene
LRBA
Gene Name
LPS responsive beige-like anchor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199282.2 9956 UTR 3 NP_001186211.2
NM_006726.4 9956 UTR 3 NP_006717.2
XM_005263372.3 9956 UTR 3 XP_005263429.1
XM_005263373.2 9956 UTR 3 XP_005263430.1
XM_005263374.3 9956 UTR 3 XP_005263431.1
XM_005263375.3 9956 UTR 3 XP_005263432.1
XM_011532434.2 9956 UTR 3 XP_011530736.1
XM_017008872.1 9956 UTR 3 XP_016864361.1
XM_017008873.1 9956 UTR 3 XP_016864362.1
XM_017008874.1 9956 UTR 3 XP_016864363.1

View Full Product Details