Product Details

SNP ID
rs12862
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:6144876 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGTTAAAGTAAGGGATATTGGCT[C/T]CTGCTGATCAGGCCACAGTGTGACA
Phenotype
MIM: 134570
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
F13A1 PubMed Links

Gene Details

Gene
F13A1
Gene Name
coagulation factor XIII A chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000129.3 3065 UTR 3 NP_000120.2

View Full Product Details