Product Details

SNP ID
rs1045343
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100706918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTAGGAAAAGGCTTCCCAGCCGC[C/T]TGCCCCGGAGACCCAATGACATTTA
Phenotype
MIM: 606113
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POP7 PubMed Links

Gene Details

Gene
POP7
Gene Name
POP7 homolog, ribonuclease P/MRP subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005837.2 350 Silent Mutation CTG,TTG L30L NP_005828.2

View Full Product Details