Product Details

SNP ID
rs803940
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:116690860 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGCCTGACAATAAATAGTAGTTG[C/T]TTTAAGAGTGTTAGCAATTATGAGA
Phenotype
MIM: 612856 MIM: 602290
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASTN2 PubMed Links

Gene Details

Gene
ASTN2
Gene Name
astrotactin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184734.1 Intron NP_001171663.1
NM_001184735.1 Intron NP_001171664.1
NM_014010.4 Intron NP_054729.3
NM_198186.3 Intron NP_937829.3
NM_198187.3 Intron NP_937830.3
NM_198188.2 Intron NP_937831.1
Gene
TRIM32
Gene Name
tripartite motif containing 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099679.1 Intron NP_001093149.1
NM_012210.3 Intron NP_036342.2
XM_005251813.3 Intron XP_005251870.1
XM_011518398.2 Intron XP_011516700.1
XM_017014486.1 Intron XP_016869975.1

View Full Product Details