Product Details

SNP ID
rs1294818
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:118507024 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TACGTGGTTCATCTTTTGTCTGTGT[A/G]CAGCCTTGCACAATCATGCCTAAAT
Phenotype
MIM: 300681
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DOCK11 PubMed Links

Gene Details

Gene
DOCK11
Gene Name
dedicator of cytokinesis 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144658.3 Intron NP_653259.3
XM_005262368.3 Intron XP_005262425.1
XM_005262369.4 Intron XP_005262426.1
XM_011531276.2 Intron XP_011529578.1
XM_011531278.2 Intron XP_011529580.1

View Full Product Details