Product Details

SNP ID
rs15487
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:92384184 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TCAAAATAAGTGAATGAGCTAATAA[A/T]CATTGTTAGAATAATAATTGTGTTC
Phenotype
MIM: 611505 MIM: 602383
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CENPP PubMed Links

Gene Details

Gene
CENPP
Gene Name
centromere protein P
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012267.2 2612 Intron NP_001012267.1
NM_001286969.1 2612 Intron NP_001273898.1
NM_001286971.1 2612 Intron NP_001273900.1
XM_011518685.2 2612 Intron XP_011516987.1
XM_011518689.1 2612 Intron XP_011516991.1
XM_017014715.1 2612 Intron XP_016870204.1
Gene
OGN
Gene Name
osteoglycin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014057.4 2612 UTR 3 NP_054776.1
NM_024416.4 2612 UTR 3 NP_077727.3
NM_033014.3 2612 UTR 3 NP_148935.1

View Full Product Details