Product Details

SNP ID
rs900056
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40239625 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCAGATATGGTGGTCAGGCTGCC[C/T]GTGTGTGTCCTCTGGAGGTGTTGGG
Phenotype
MIM: 608110
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BUB1B-PAK6 PubMed Links

Gene Details

Gene
BUB1B-PAK6
Gene Name
BUB1B-PAK6 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128628.2 535 Intron NP_001122100.1
NM_001128629.2 535 Intron NP_001122101.1
Gene
PAK6
Gene Name
p21 (RAC1) activated kinase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276717.1 535 Intron NP_001263646.1
NM_001276718.1 535 UTR 5 NP_001263647.1
NM_020168.5 535 Intron NP_064553.1

View Full Product Details