Product Details

SNP ID
rs896374
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22442234 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTATGTGCTGCTTTTATTCAGC[C/G]GTAAGTCTTAGGCACAACCCTTCTC
Phenotype
MIM: 114107 MIM: 608736
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PPP3CC PubMed Links

Gene Details

Gene
PPP3CC
Gene Name
protein phosphatase 3 catalytic subunit gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243974.1 Intron NP_001230903.1
NM_001243975.1 Intron NP_001230904.1
NM_005605.4 Intron NP_005596.2
XM_017013610.1 Intron XP_016869099.1
XM_017013611.1 Intron XP_016869100.1
Gene
SLC39A14
Gene Name
solute carrier family 39 member 14
There are no transcripts associated with this gene.

View Full Product Details