Product Details

SNP ID
rs1757456
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41501296 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGCGCCTGTATCAGCACCCGCCAC[A/T]CAGGCGCACAAATAAACCTTCCCAG
Phenotype
MIM: 147521 MIM: 151520
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ITPKA PubMed Links

Gene Details

Gene
ITPKA
Gene Name
inositol-trisphosphate 3-kinase A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002220.2 170 Intron NP_002211.1
XM_011521522.2 170 UTR 5 XP_011519824.1
Gene
LTK
Gene Name
leukocyte receptor tyrosine kinase
There are no transcripts associated with this gene.

View Full Product Details