Product Details

SNP ID
rs1706343
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:57378079 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAACACAGTGTATAACATGGCACA[C/T]CTGAGATGACACCTGAGACACCTGT
Phenotype
MIM: 607856
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CGNL1 PubMed Links

Gene Details

Gene
CGNL1
Gene Name
cingulin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252335.1 Intron NP_001239264.1
NM_032866.4 Intron NP_116255.2
XM_005254726.3 Intron XP_005254783.1
XM_005254727.4 Intron XP_005254784.1
XM_011522120.1 Intron XP_011520422.1
XM_011522121.1 Intron XP_011520423.1
XM_017022685.1 Intron XP_016878174.1
XM_017022686.1 Intron XP_016878175.1

View Full Product Details