Product Details

SNP ID
rs1139701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:58567223 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCACCCACCTGGTTCAGCCTCTG[C/T]TGCCGTCTCAGCAGCTCCTGCTCAA
Phenotype
MIM: 608295
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM107A PubMed Links

Gene Details

Gene
FAM107A
Gene Name
family with sequence similarity 107 member A
There are no transcripts associated with this gene.

View Full Product Details