Product Details

SNP ID
rs1145086
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.15:45362129 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCTGTTCAGTCCAAGTAGGACTGTA[A/G]GGTGCCTCGGCGCCGGACATCGCAG
Phenotype
MIM: 602360
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GATM PubMed Links

Gene Details

Gene
GATM
Gene Name
glycine amidinotransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321015.1 1581 Silent Mutation ATA,GTA I289V NP_001307944.1
NM_001482.2 1581 Silent Mutation ATA,GTA I418V NP_001473.1

View Full Product Details