Product Details

SNP ID
rs1045118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:64516286 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAATATCACTCTGTAATGAAGCTG[A/C]TTGGGGGATCCTCCATTTGATTGTG
Phenotype
MIM: 605421
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADAMTS9 PubMed Links

Gene Details

Gene
ADAMTS9
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318781.1 6681 UTR 3 NP_001305710.1
NM_182920.1 6681 UTR 3 NP_891550.1

View Full Product Details