Product Details

SNP ID
rs1045730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:65355652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGATGATCACGAACCTGACTGTCT[G/T]TGCTTGGGCTGTATTTGAGTAAAAG
Phenotype
MIM: 602625
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MAGI1 PubMed Links

Gene Details

Gene
MAGI1
Gene Name
membrane associated guanylate kinase, WW and PDZ domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033057.1 5822 UTR 3 NP_001028229.1
NM_004742.2 5822 Intron NP_004733.2
NM_015520.1 5822 UTR 3 NP_056335.1
XM_005265563.1 5822 UTR 3 XP_005265620.1
XM_005265564.1 5822 UTR 3 XP_005265621.1
XM_005265565.1 5822 UTR 3 XP_005265622.1
XM_005265566.1 5822 UTR 3 XP_005265623.1
XM_005265568.1 5822 UTR 3 XP_005265625.1
XM_005265570.1 5822 UTR 3 XP_005265627.1
XM_005265571.1 5822 UTR 3 XP_005265628.1
XM_005265574.3 5822 UTR 3 XP_005265631.1
XM_006713407.1 5822 UTR 3 XP_006713470.1
XM_006713408.1 5822 UTR 3 XP_006713471.1
XM_006713409.1 5822 UTR 3 XP_006713472.1
XM_006713410.1 5822 UTR 3 XP_006713473.1
XM_006713411.1 5822 UTR 3 XP_006713474.1
XM_006713412.1 5822 UTR 3 XP_006713475.1
XM_006713414.3 5822 UTR 3 XP_006713477.1
XM_011534236.1 5822 UTR 3 XP_011532538.1
XM_011534240.2 5822 UTR 3 XP_011532542.1
XM_011534241.2 5822 UTR 3 XP_011532543.1
XM_017007497.1 5822 UTR 3 XP_016862986.1
XM_017007498.1 5822 UTR 3 XP_016862987.1
XM_017007499.1 5822 UTR 3 XP_016862988.1
XM_017007500.1 5822 UTR 3 XP_016862989.1
XM_017007501.1 5822 UTR 3 XP_016862990.1
XM_017007502.1 5822 UTR 3 XP_016862991.1
XM_017007503.1 5822 UTR 3 XP_016862992.1
XM_017007504.1 5822 UTR 3 XP_016862993.1
XM_017007505.1 5822 UTR 3 XP_016862994.1
XM_017007506.1 5822 UTR 3 XP_016862995.1
XM_017007507.1 5822 UTR 3 XP_016862996.1
XM_017007508.1 5822 UTR 3 XP_016862997.1

View Full Product Details