Product Details

SNP ID
rs1399570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:157413636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTACACTTCTGTATTAAAATCACA[A/C]GAAAAGCAAATGATCTATGGTCTGC
Phenotype
MIM: 604448
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CYTIP PubMed Links

Gene Details

Gene
CYTIP
Gene Name
cytohesin 1 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004288.4 3384 Intron NP_004279.3
XM_017005386.1 3384 UTR 3 XP_016860875.1

View Full Product Details