Product Details

SNP ID
rs1263597
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:207082161 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGCTATTTGATATTCTCTTGGGTA[C/G]CAACCCTTCTGCATTCTCATTTGTG
Phenotype
MIM: 604865
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
KLF7 PubMed Links

Gene Details

Gene
KLF7
Gene Name
Kruppel like factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270942.1 Intron NP_001257871.1
NM_001270943.1 Intron NP_001257872.1
NM_001270944.1 Intron NP_001257873.1
NM_003709.3 Intron NP_003700.1
XM_005246926.3 Intron XP_005246983.1
XM_006712815.3 Intron XP_006712878.1
XM_011512072.2 Intron XP_011510374.1
XM_011512073.2 Intron XP_011510375.1
XM_011512074.2 Intron XP_011510376.1
XM_011512075.1 Intron XP_011510377.1
XM_017005161.1 Intron XP_016860650.1
XM_017005162.1 Intron XP_016860651.1
XM_017005163.1 Intron XP_016860652.1
XM_017005164.1 Intron XP_016860653.1

View Full Product Details