Product Details

SNP ID
rs1345827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:35904787 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATGGCTGAGGAACTGATCTAAAA[A/C]AAAAGAACAAGTGAAGCTGAGGATG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CAPSL PubMed Links

Gene Details

Gene
CAPSL
Gene Name
calcyphosine like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042625.1 Intron NP_001036090.1
NM_144647.3 Intron NP_653248.3
XM_006714444.3 Intron XP_006714507.1
XM_006714445.3 Intron XP_006714508.1

View Full Product Details