Product Details

SNP ID
rs885291
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:219912128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACACCTGTCTCTACTGAACTTTA[A/G]AGTTCTTGATTTACCACCACTGGAA
Phenotype
MIM: 611146
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC30A10 PubMed Links

Gene Details

Gene
SLC30A10
Gene Name
solute carrier family 30 member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018713.2 5326 Intron NP_061183.2
XM_006711437.3 5326 UTR 3 XP_006711500.2
XM_017001684.1 5326 UTR 3 XP_016857173.1

View Full Product Details