Product Details

SNP ID
rs844544
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196743980 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATTACTTTTGGATGAATAAAAT[A/G]TAATTTTTCAGGAGAAAAAATATGT
Phenotype
MIM: 605022 MIM: 610276
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAK2 PubMed Links

Gene Details

Gene
PAK2
Gene Name
p21 (RAC1) activated kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002577.4 Intron NP_002568.2
XM_011512870.2 Intron XP_011511172.1
XM_017006501.1 Intron XP_016861990.1
Gene
PIGX
Gene Name
phosphatidylinositol glycan anchor biosynthesis class X
There are no transcripts associated with this gene.

View Full Product Details