Product Details

SNP ID
rs1152188
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:70259053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCACATAGTGGTCAAACTCTTCA[A/G]GCCGAGTGGTGTCATGGCACAGGAC
Phenotype
MIM: 301770 MIM: 300038
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARR3 PubMed Links

Gene Details

Gene
ARR3
Gene Name
arrestin 3
There are no transcripts associated with this gene.

Gene
P2RY4
Gene Name
pyrimidinergic receptor P2Y4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002565.3 752 Missense Mutation NP_002556.1

View Full Product Details