Product Details

SNP ID
rs901740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176642830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCAGCCTGGCCTCTCTACCCCCA[G/A]TGCACCATGGCCCCGCCCTCTCCCC
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
EIF4E1B PubMed Links

Gene Details

Gene
EIF4E1B
Gene Name
eukaryotic translation initiation factor 4E family member 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099408.1 Intron NP_001092878.1
XM_011534499.2 Intron XP_011532801.1
Gene
TSPAN17
Gene Name
tetraspanin 17
There are no transcripts associated with this gene.

View Full Product Details