Product Details

SNP ID
rs1045031
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:44844689 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GACATCTATGAGAAGGACCTACGTA[C/T]GATGATATCAATGTAAGCCATCCCG
Phenotype
MIM: 300678
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP21 PubMed Links

Gene Details

Gene
DUSP21
Gene Name
dual specificity phosphatase 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022076.3 687 Missense Mutation ACG,ATG T186M NP_071359.3

View Full Product Details