Product Details

SNP ID
rs1059302
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:172210343 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACTTCACATCCAAACAGGTGTG[C/G]AGGGCTGACATTTTCCAACTGAGAA
Phenotype
MIM: 610174
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EFCAB9 PubMed Links

Gene Details

Gene
EFCAB9
Gene Name
EF-hand calcium binding domain 9
There are no transcripts associated with this gene.

Gene
UBTD2
Gene Name
ubiquitin domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152277.2 2318 UTR 3 NP_689490.2
XM_017010022.1 2318 UTR 3 XP_016865511.1
XM_017010023.1 2318 UTR 3 XP_016865512.1

View Full Product Details