Product Details

SNP ID
rs1177579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:126708441 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCAGGGCTGTGAACATGTTATT[C/T]TGGAGGCCACGCTGCAGTTCTCTGG
Phenotype
MIM: 615634
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHCHD6 PubMed Links

Gene Details

Gene
CHCHD6
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320610.1 Intron NP_001307539.1
NM_032343.2 Intron NP_115719.1

View Full Product Details