Product Details

SNP ID
rs1050525
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:5119682 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAAGAGACGTGGGACGAGTCCAT[G/T]CTCTGCAGGTTTACACCGCTGGAGC
Phenotype
MIM: 603549 MIM: 176740 MIM: 617019
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CDS2 PubMed Links

Gene Details

Gene
CDS2
Gene Name
CDP-diacylglycerol synthase 2
There are no transcripts associated with this gene.

Gene
PCNA
Gene Name
proliferating cell nuclear antigen
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002592.2 356 Missense Mutation AGA,AGC R39S NP_002583.1
NM_182649.1 356 Intron NP_872590.1
Gene
PCNA-AS1
Gene Name
PCNA antisense RNA 1
There are no transcripts associated with this gene.

Gene
TMEM230
Gene Name
transmembrane protein 230
There are no transcripts associated with this gene.

View Full Product Details