Product Details

SNP ID
rs801629
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:46769546 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCATGTCCTCCTTACACAGGTCCCG[C/T]TTCCGCCGAGACCTGGCTGTACAGC
Phenotype
MIM: 616879
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TBC1D22A PubMed Links

Gene Details

Gene
TBC1D22A
Gene Name
TBC1 domain family member 22A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284303.1 Intron NP_001271232.1
NM_001284304.1 Intron NP_001271233.1
NM_001284305.1 Intron NP_001271234.1
NM_014346.3 Intron NP_055161.1
XM_005261496.1 Intron XP_005261553.1
XM_011530099.1 Intron XP_011528401.1
XM_011530100.2 Intron XP_011528402.1
XM_011530101.2 Intron XP_011528403.1
XM_011530102.2 Intron XP_011528404.1
XM_017028742.1 Intron XP_016884231.1
XM_017028743.1 Intron XP_016884232.1
XM_017028744.1 Intron XP_016884233.1
XM_017028745.1 Intron XP_016884234.1
XM_017028746.1 Intron XP_016884235.1
XM_017028747.1 Intron XP_016884236.1

View Full Product Details