Product Details

SNP ID
rs1144833
Assay Type
Validated
NCBI dbSNP Submissions
51
Location
Chr.1:226657368 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
TTTACATTCTCTTCTTAGCCTCTTT[T/C]TCACCTGACTTGTTCCCTCAATTTG
Phenotype
MIM: 147522
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
ITPKB PubMed Links

Gene Details

Gene
ITPKB
Gene Name
inositol-trisphosphate 3-kinase B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002221.3 Intron NP_002212.3
XM_005273120.2 Intron XP_005273177.1
XM_017001211.1 Intron XP_016856700.1
Gene
ITPKB-IT1
Gene Name
ITPKB intronic transcript 1
There are no transcripts associated with this gene.

View Full Product Details