Product Details

SNP ID
rs1370389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:64093101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACTTATAATACAAGTTTCCTGAC[A/C]TGCATTTCCTGAGTGAACCCAAATG
Phenotype
MIM: 614797
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PELI1 PubMed Links

Gene Details

Gene
PELI1
Gene Name
pellino E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020651.3 3977 UTR 3 NP_065702.2
XM_005264437.3 3977 UTR 3 XP_005264494.1
XM_011532994.2 3977 UTR 3 XP_011531296.1
XM_011532996.2 3977 UTR 3 XP_011531298.1
XM_011532997.2 3977 UTR 3 XP_011531299.1
XM_017004520.1 3977 UTR 3 XP_016860009.1
XM_017004521.1 3977 UTR 3 XP_016860010.1

View Full Product Details