Product Details

SNP ID
rs698910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:207765913 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATAAAGGGGGAGGGGGGAGGGG[G/T]GAGGGGGTGGAGCAAATAAGTGTTG
Phenotype
MIM: 601723
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCNYL1 PubMed Links

Gene Details

Gene
CCNYL1
Gene Name
cyclin Y like 1
There are no transcripts associated with this gene.

Gene
FZD5
Gene Name
frizzled class receptor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003468.3 3237 UTR 3 NP_003459.2

View Full Product Details