Product Details

SNP ID
rs1063638
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:38797868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTTTTTCCCCTTCTCTGTTCTG[A/T]CTTCTCTCAGTCGAGCCTTGGCGAC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DHX57 PubMed Links

Gene Details

Gene
DHX57
Gene Name
DEAH-box helicase 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011533154.2 4695 UTR 3 XP_011531456.1
XM_011533155.2 4695 UTR 3 XP_011531457.1
XM_011533156.2 4695 Intron XP_011531458.1
XM_011533157.1 4695 Intron XP_011531459.1

View Full Product Details