Product Details

SNP ID
rs14294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:65311953 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGGAAAGGGAGTGGGGAGCAGGCC[A/G]TGTCTTCTGCTGGCCGCTACCACAG
Phenotype
MIM: 609292
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPRED2 PubMed Links

Gene Details

Gene
SPRED2
Gene Name
sprouty related EVH1 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128210.1 2755 UTR 3 NP_001121682.1
NM_181784.2 2755 UTR 3 NP_861449.2
XM_005264200.4 2755 Intron XP_005264257.2
XM_005264201.1 2755 UTR 3 XP_005264258.1
XM_005264202.4 2755 Intron XP_005264259.1
XM_017003576.1 2755 UTR 3 XP_016859065.1

View Full Product Details