Product Details

SNP ID
rs859030
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18792340 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTTAGGGTAGTCACATAGCAAG[C/T]ACCTAGTAACTATGAGAGAAATGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00652 PubMed Links

Gene Details

Gene
LINC00652
Gene Name
long intergenic non-protein coding RNA 652
There are no transcripts associated with this gene.

Gene
LOC100270804
Gene Name
uncharacterized LOC100270804
There are no transcripts associated with this gene.

View Full Product Details